Learn More
Sklar Instruments Scissors, Sklar Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Straight, Razor, Stainless Steel, EA.
SDP
Catalog No. 501093417 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklarhone Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Razor, Stainless Steel, EA. Sklarhone Iris Scissors feature one knife-like razor sharp edge and one standard scissor edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone premium scissors. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Sklarhone instruments are identified by dual black finger rings. This product is straight with one razor edge blade, sharp tips, and an overall length of 4-1/2 inches.
Catalog No. 50-109-3417 Supplier Sklar Instruments Supplier No. 153352
Scissors, Sklarhone Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Razor, Stainless Steel, EA. Sklarhone Iris Scissors feature one knife-like razor sharp edge and one standard scissor edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone premium scissors. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Sklarhone instruments are identified by dual black finger rings. This product is straight with one razor edge blade, sharp tips, and an overall length of 4-1/2 inches.