Learn More
Sklar Instruments Scissors, Stevens Tenotomy Scissors, Sklar 4-1/2 in., Blunt/Blunt, Curved, Smooth, Extra Light (Sklar), Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 231157
Encompass_Preferred
Scissors, Stevens Tenotomy Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Curved, Smooth, Extra Light (SklarLite), Stainless Steel, EA. Sklarlite Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with variable tip styles. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Stevens tenotomy scissors are available straight or curved and with sharp or blunt tips, in order to accommodate different procedures. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.
Catalog No. 50-109-3683