Learn More
Sklar Instruments Scissors, Stevens Tenotomy Scissors, Sklar 4-1/2 in., Blunt/Blunt, Straight, Smooth, Delicate, Stainless Steel, EA. Sklarvens Tenotomy Scissors are used for delicate dissection.
SDP
Catalog No. 501093924 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Stevens Tenotomy Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have a firm grip and control while dissecting or cutting in small or narrow areas.
Catalog No. 50-109-3924 Supplier Sklar Instruments Supplier No. 641242
Scissors, Stevens Tenotomy Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have a firm grip and control while dissecting or cutting in small or narrow areas.