Learn More
Sklar Instruments Scissors,Surgi-OR Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094026 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors,Surgi-OR Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Hand-crafted in Pakistan, Surgi-OR Iris Scissors are a mid-grade instrument forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth, sharp/sharp tips and 4-1/2 inches in length.
Catalog No. 50-109-4026 Supplier Sklar Instruments Supplier No. 95109
Scissors,Surgi-OR Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Hand-crafted in Pakistan, Surgi-OR Iris Scissors are a mid-grade instrument forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth, sharp/sharp tips and 4-1/2 inches in length.