Learn More
Sklar Instruments Scissors,Surgi-OR Spencer Stitch Scissors, Sklar 4-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094074 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors,Surgi-OR Spencer Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Surgi-OR Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Catalog No. 50-109-4074 Supplier Sklar Instruments Supplier No. 95352
Scissors,Surgi-OR Spencer Stitch Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Surgi-OR Spencer Stitch Scissors are primarily used for suture removal. These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. They are most commonly available in lengths of 3-1/2 in. and 4-1/2 in. and therefore typically have smaller, more delicate tips that are used for removing smaller sutures. They should not be used for cutting tissue. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.