Learn More
Sklar Instruments Scissors, Wagner Scissors, Sklar 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501093813 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Wagner Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA.Sklar's Wagner Scissors are finger ring surgical scissors most commonly used in procedures requiring delicate or precise cutting, particularly plastic surgery. Curvature, blade, and tip style can be tailored to meet the needs of specific procedures. The standard length for Wagner scissors is 4-1/2 inches. This product is curved with smooth blades and sharp/sharp tips.
Catalog No. 50-109-3813 Supplier Sklar Instruments Supplier No. 471058
Scissors, Wagner Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel, EA.Sklar's Wagner Scissors are finger ring surgical scissors most commonly used in procedures requiring delicate or precise cutting, particularly plastic surgery. Curvature, blade, and tip style can be tailored to meet the needs of specific procedures. The standard length for Wagner scissors is 4-1/2 inches. This product is curved with smooth blades and sharp/sharp tips.