Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1,3-Oxazol-5-yl)aniline, 97%, Thermo Scientific™

Catalog No. DFP00120DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
DFP00120DA 1 g Amber glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DFP00120DA Supplier Thermo Scientific Chemicals Supplier No. DFP00120DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1008-95-3
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.176
MDL Number MFCD00052192
InChI Key SLJBMRSOKUTXDF-UHFFFAOYSA-N
PubChem CID 517747
IUPAC Name 4-(1,3-oxazol-5-yl)aniline
SMILES C1=CC(=CC=C1C2=CN=CO2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4-1,3-oxazol-5-yl aniline, 4-5-oxazolyl aniline, 4-oxazol-5-yl aniline, benzenamine, 4-5-oxazolyl, 5-4-aminophenyl oxazole, benzenamine,4-5-oxazolyl, 4-oxazol-5-yl-aniline, maybridge3_001299, 4-oxazol-5-yl-phenylamine
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.