Learn More
Sklar Instruments Scissors, Dissecting Scissors, Sklar 4 -1/3 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501093351 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Dissecting Scissors, Sklar, 4-1/3 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar Dissecting Scissors are used primarily for separating tissue or cutting specimen for study. These standard scissors should be used with tissue that is neither too delicate nor too tough, upon which tailored dissecting scissors, such as Mayo or Metzenbaum, would be more appropriate. This product is straight with smooth edges, sharp/sharp tips, and an overall length of 4-1/3 inches.
Catalog No. 50-109-3351 Supplier Sklar Instruments Supplier No. 151270
Scissors, Dissecting Scissors, Sklar, 4-1/3 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Sklar Dissecting Scissors are used primarily for separating tissue or cutting specimen for study. These standard scissors should be used with tissue that is neither too delicate nor too tough, upon which tailored dissecting scissors, such as Mayo or Metzenbaum, would be more appropriate. This product is straight with smooth edges, sharp/sharp tips, and an overall length of 4-1/3 inches.