Learn More
Medchemexpress LLC 4-[1-(4-cyclohexylphenyl)-3-(3-methanesulfonylphenyl)ureidomethyl]-N-(2H-tetrazol-5-yl)benzamide | 307986-98-7 | 98.4% | 573.67 g·mol⁻1 | C29H31N7O4S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12462250MG
Encompass_Preferred
NNC-0640 is a small-molecule negative allosteric modulator (NAM) of the human glucagon receptor (GCGR) used for biochemical and preclinical research. It has an IC50 of 69.2 nM against human GCGR, a molecular weight of 573.67 g·mol⁻1, formula C29H31N7O4S, and supplier-reported purity >98%.
  • Negative allosteric modulator of human glucagon receptor with IC50 69.2 nM.
  • High purity suitable for biochemical and preclinical research (>98%).
  • Characterized molecular weight 573.67 g·mol⁻1 and formula C29H31N7O4S.
  • Available as solid and as a 10 mM solution in DMSO for assay use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-36275