Learn More
Sklar Instruments Scissors, Sealy Dissecting Scissors, Sklar 4-1/4 in., Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501093978 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sealy Dissecting Scissors, Sklar, 4-1/4 in., Straight, Smooth, Stainless Steel, EA. Sklar's Sealy Dissecting Scissors are finger ring scissors used for dissecting tissue. This product has straight, blunt/blunt tips, and an overall length of 4-1/4 inches.
Catalog No. 50-109-3978 Supplier Sklar Instruments Supplier No. 643285
Scissors, Sealy Dissecting Scissors, Sklar, 4-1/4 in., Straight, Smooth, Stainless Steel, EA. Sklar's Sealy Dissecting Scissors are finger ring scissors used for dissecting tissue. This product has straight, blunt/blunt tips, and an overall length of 4-1/4 inches.