Learn More
Sklar Instruments Surgical Scrub Brush, Sklar 4-1/4 in., Nylon / Plastic, Disposable, EA.
SDP
Catalog No. 501094459 Shop All Sklar Instruments Products
Encompass_Preferred
Surgical Scrub Brush, Sklar, 4-1/4 in., Nylon / Plastic, Disposable, EA. Sklar's Surgical Scrub Brush with Nail Scrubber features hard nylon bristles used to remove debris and microorganisms from the hands, nails, and forearms in order to reduce and prevent the spread of infectious diseases.
Catalog No. 50-109-4459 Supplier Sklar Instruments Supplier No. 062882
Surgical Scrub Brush, Sklar, 4-1/4 in., Nylon / Plastic, Disposable, EA. Sklar's Surgical Scrub Brush with Nail Scrubber features hard nylon bristles used to remove debris and microorganisms from the hands, nails, and forearms in order to reduce and prevent the spread of infectious diseases.