Learn More
Arrow Engineering Type 316 Stainless Steel VPP Series Propeller 4-1/4"dia. 3 Blade
Catalog No. NC3397181
Encompass_Preferred
The Arrow Mixing Products Type 316 Stainless Steel VPP Series 4-1/4″dia. 3 blade propeller with 5/16-24 tapped hole. Our stainless propeller selection is engineered for optimal performance, ensuring smooth and consistent mixing for various industries, including pharmaceuticals, food processing, and chemical production. With a carefully crafted design, these propellers maximize power and efficiency, helping you achieve the best results for your mixing needs.
Catalog No. NC3397181 Supplier Arrow Engineering Supplier No. VPP31642
The Arrow Mixing Products Type 316 Stainless Steel VPP Series 4-1/4″dia. 3 blade propeller with 5/16-24 tapped hole. Our stainless propeller selection is engineered for optimal performance, ensuring smooth and consistent mixing for various industries, including pharmaceuticals, food processing, and chemical production. With a carefully crafted design, these propellers maximize power and efficiency, helping you achieve the best results for your mixing needs.