Learn More
Sklar Instruments Scissors, Surgi-OR Stevens Tenotomy Scissors, Sklar 4-1/8 in., Blunt/Blunt, Tenotomy Scissors, Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094045 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Surgi-OR Stevens Tenotomy Scissors, Sklar, 4-1/8 in., Blunt/Blunt, Tenotomy Scissors, Curved, Smooth, Stainless Steel, EA. Surgi-OR Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Stevens tenotomy scissors are available straight or curved and with sharp or blunt tips, in order to accommodate different procedures. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.
Catalog No. 50-109-4045 Supplier Sklar Instruments Supplier No. 95161
Scissors, Surgi-OR Stevens Tenotomy Scissors, Sklar, 4-1/8 in., Blunt/Blunt, Tenotomy Scissors, Curved, Smooth, Stainless Steel, EA. Surgi-OR Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Stevens tenotomy scissors are available straight or curved and with sharp or blunt tips, in order to accommodate different procedures. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.