Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific™

Catalog No. AAH6370022
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6370022 100 g
AAH6370006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6370022 Supplier Thermo Scientific Chemicals Supplier No. H6370022

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134855-87-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00143209
InChI Key CDQPLIAKRDYOCB-UHFFFAOYSA-N
Synonym 4-1-aminoethyl phenol, 1-p-hydroxyphenyl ethylamine, phenol, 4-1-aminoethyl, 4-aminoethyl phenol, 1-4-hydroxyphenyl ethylamine, p-oxyphenylathylamin, timtec-bb sbb008453, 4-1-aminoethyl-pheno, 4-1-amino-ethyl-phenol, 1-4-hydroxyphenyl ethanamine
PubChem CID 123594
ChEBI CHEBI:581
IUPAC Name 4-(1-aminoethyl)phenol
SMILES CC(C1=CC=C(C=C1)O)N

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity 97%
Chemical Name or Material (+/-)-4-(1-Aminoethyl)phenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber SJ5950150
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.