Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[(1-methyl-1H-pyrrol-2-yl)methylene]-1,3(2H,4H)-isoquinolinedione | 1104546-89-5 | 98.7% | 252.27 g/mol | C15H12N2O2 | 5 MG
SDP

Catalog No. 5000263405
Encompass_Preferred

BYK204165 is a small-molecule PARP1 inhibitor for research use (CAS 1104546-89-5). It has molecular formula C15H12N2O2, molecular weight 252.27 g/mol, and is provided at high purity for use in biochemical and cell-based assays.

  • High purity suitable for biochemical assays.
  • Molecular weight 252.27 g/mol.
  • Chemical formula C15H12N2O2.
  • Provided in milligram-scale quantities for laboratory research.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-63405 Supplier Medchemexpress LLC Supplier No. HY1086325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BYK204165 is a small-molecule PARP1 inhibitor for research use (CAS 1104546-89-5). It has molecular formula C15H12N2O2, molecular weight 252.27 g/mol, and is provided at high purity for use in biochemical and cell-based assays.

  • High purity suitable for biochemical assays.
  • Molecular weight 252.27 g/mol.
  • Chemical formula C15H12N2O2.
  • Provided in milligram-scale quantities for laboratory research.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.