Learn More
Medchemexpress LLC 4-(1-methylethyl)phenyl diphenyl phosphate | 55864-04-5 | MFCD00377267 | 368.36 | C21H21O4P | 50 MG
SDP
Catalog No. 5000284523
Encompass_Preferred
4-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is described by the manufacturer with key chemical identifiers and handling guidance.
  • Organophosphate flame retardant used in materials research
  • Chemical identifiers: CAS 55864-04-5; formula C21H21O4P
  • Molecular weight 368.36 g·mol⁻¹
  • Available in small research pack sizes, for example 50 MG
  • Supplied for research use only; not for human or clinical use
  • Refer to the Certificate of Analysis and SDS for storage and handling instructions
Catalog No. 50-002-84523 Supplier Medchemexpress LLC Supplier No. HYW58826850MG
4-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is described by the manufacturer with key chemical identifiers and handling guidance.
  • Organophosphate flame retardant used in materials research
  • Chemical identifiers: CAS 55864-04-5; formula C21H21O4P
  • Molecular weight 368.36 g·mol⁻¹
  • Available in small research pack sizes, for example 50 MG
  • Supplied for research use only; not for human or clinical use
  • Refer to the Certificate of Analysis and SDS for storage and handling instructions