Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(1-methylethyl)phenyl diphenyl phosphate | 55864-04-5 | MFCD00377267 | 95.0% | 368.36 g/mol | C21H21O4P | 100 MG
SDP

Catalog No. 5000284582
Encompass_Preferred

4-(1-methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. Molecular formula C21H21O4P and molecular weight 368.36 g/mol. The material is provided for biochemical assay and research applications; store under the conditions specified in the product Certificate of Analysis.

  • Organophosphate flame retardant
  • Intended for research use only
  • Molecular formula: C21H21O4P
  • Molecular weight: 368.36 g/mol
  • CAS number: 55864-04-5
  • Follow storage conditions in the Certificate of Analysis

Catalog No. 50-002-84582 Supplier Medchemexpress LLC Supplier No. HYW588268100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(1-methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. Molecular formula C21H21O4P and molecular weight 368.36 g/mol. The material is provided for biochemical assay and research applications; store under the conditions specified in the product Certificate of Analysis.

  • Organophosphate flame retardant
  • Intended for research use only
  • Molecular formula: C21H21O4P
  • Molecular weight: 368.36 g/mol
  • CAS number: 55864-04-5
  • Follow storage conditions in the Certificate of Analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.