Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(1-methylethyl)phenyl diphenyl phosphate | 55864-04-5 | MFCD00377267 | 98.0% | 368.36 g/mol | C21H21O4P | 5 MG
SDP

Catalog No. 5000284524
Encompass_Preferred

4-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied as a research reagent for laboratory and materials science applications. It is used as an additive to impart flame resistance to polymers and as a reagent for analytical and materials testing. Typical identifiers include CAS 55864-04-5, formula C21H21O4P, and molecular weight 368.36 g/mol.

  • Acts as a flame retardant additive for polymers and materials.
  • Suitable for laboratory research and analytical applications.
  • Available in small-quantity packaging for research-scale use.
  • Chemical formula C21H21O4P and molecular weight 368.36 g/mol.
  • CAS number 55864-04-5 for unambiguous identification.
  • Purity typically reported around 95-98% depending on vendor and lot.

Catalog No. 50-002-84524 Supplier Medchemexpress LLC Supplier No. HYW5882685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied as a research reagent for laboratory and materials science applications. It is used as an additive to impart flame resistance to polymers and as a reagent for analytical and materials testing. Typical identifiers include CAS 55864-04-5, formula C21H21O4P, and molecular weight 368.36 g/mol.

  • Acts as a flame retardant additive for polymers and materials.
  • Suitable for laboratory research and analytical applications.
  • Available in small-quantity packaging for research-scale use.
  • Chemical formula C21H21O4P and molecular weight 368.36 g/mol.
  • CAS number 55864-04-5 for unambiguous identification.
  • Purity typically reported around 95-98% depending on vendor and lot.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.