Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. N07981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07981G 1 g
N0798-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07981G Supplier TCI America Supplier No. N07981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 870774-25-7
Molecular Formula C16H13BO2
InChI Key BQHVXFQXTOIMQM-UHFFFAOYSA-N
PubChem CID 44119399

Specifications

Color White-Yellow
Quantity 1 g
Synonym 4-naphthalen-1-yl phenylboronic acid, 4-1-naphthyl phenylboronic acid, 4-naphthalen-1-yl phenyl boronic acid, 4-1-naphthyl benzeneboronic acid, 4-naphthalen-1-yl phenylboronicacid, boronic acid, 4-1-naphthalenyl phenyl, 4-1-naphthyl phenyl boronic acid
SMILES B(C1=CC=C(C=C1)C2=CC=CC3=CC=CC=C32)(O)O
IUPAC Name (4-naphthalen-1-ylphenyl)boronic acid
Molecular Weight (g/mol) 248.088
Formula Weight 248.09
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Naphthyl)phenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.