Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(1-phenylethyl)resorcinol | 85-27-8 | MFCD21648466 | 99.4% | 214.26 g·mol⁻¹ | C14H14O2 | 25G
SDP

Catalog No. 5000236458
Encompass_Preferred

4-(1-Phenylethyl)resorcinol (CAS 85-27-8) is a phenolic resorcinol derivative used as a skin-lightening active in cosmetic research and as a biochemical reagent for studies of melanogenesis. It inhibits melanin production to help reduce hyperpigmentation and dark spots, and is supplied as a high-purity solid suitable for formulation and analytical applications.

  • Used as a skin-lightening active and biochemical reagent.
  • Inhibits melanin synthesis to reduce hyperpigmentation and dark spots.
  • Molecular formula C14H14O2 and molecular weight 214.26 g·mol⁻¹.
  • High purity (99.4%) suitable for formulation and analytical use.
  • Off-white to yellow solid appearance.
  • Available in laboratory packaging, including 25 g quantity.

Catalog No. 50-002-36458 Supplier Medchemexpress LLC Supplier No. HY12580925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-(1-Phenylethyl)resorcinol (CAS 85-27-8) is a phenolic resorcinol derivative used as a skin-lightening active in cosmetic research and as a biochemical reagent for studies of melanogenesis. It inhibits melanin production to help reduce hyperpigmentation and dark spots, and is supplied as a high-purity solid suitable for formulation and analytical applications.

  • Used as a skin-lightening active and biochemical reagent.
  • Inhibits melanin synthesis to reduce hyperpigmentation and dark spots.
  • Molecular formula C14H14O2 and molecular weight 214.26 g·mol⁻¹.
  • High purity (99.4%) suitable for formulation and analytical use.
  • Off-white to yellow solid appearance.
  • Available in laboratory packaging, including 25 g quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.