Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Propenyl)-1,2-dimethoxybenzene 98.0+%, TCI America™
SDP

Catalog No. P110325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P110325ML 25 mL
P1103500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P110325ML Supplier TCI America Supplier No. P110325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93-16-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009282
InChI Key NNWHUJCUHAELCL-PLNGDYQASA-N
Synonym 1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether
PubChem CID 1549045
ChEBI CHEBI:50550
IUPAC Name 1,2-dimethoxy-4-[(Z)-prop-1-enyl]benzene
SMILES CC=CC1=CC(=C(C=C1)OC)OC

Specifications

Color Yellow
Boiling Point 264°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(1-Propenyl)-1,2-dimethoxybenzene
EINECSNumber (3)-0638
RTECSNumber CZ7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.