Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Pyrrolidinyl)benzonitrile, ≥90%, Thermo Scientific™

Catalog No. CC30116EB
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30116EB 25 g
CC30116DA 1 g
CC30116EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC30116EB Supplier Thermo Scientific Chemicals Supplier No. CC30116EB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10282-30-1
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD07368512
InChI Key ZNMSYUCZLWETII-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
PubChem CID 4961271
IUPAC Name 4-pyrrolidin-1-ylbenzonitrile
SMILES C1CCN(C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 25 g
Formula Weight 172.23
Percent Purity 90%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.