Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Pyrrolidinyl)benzonitrile 96.0+%, TCI America™
SDP

Catalog No. P24281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24281G 1 g
P24285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24281G Supplier TCI America Supplier No. P24281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10282-30-1
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD07368512
InChI Key ZNMSYUCZLWETII-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
PubChem CID 4961271
IUPAC Name 4-pyrrolidin-1-ylbenzonitrile
SMILES C1CCN(C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 172.23
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Pyrrolidinyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.