Learn More
Bio Rad Laboratories 4-12% BIS-TRIS GEL, 26W
Catalog No. NC9081977
Encompass
Criterion XT Bis-Tris Gel, 4 to 12%, 26-well, 15microliter, 13.3cm x 8.7cm, Solid, White, Odorless, Boiling point: 100degreeC, Flash point: >100degreeC, pH: 6.4 at 20degreeC, Ignition temperature: 400.0degreeC
Catalog No. NC9081977 Supplier Bio Rad Laboratories Supplier No. 3450125
Criterion XT Bis-Tris Gel, 4 to 12%, 26-well, 15microliter, 13.3cm x 8.7cm, Solid, White, Odorless, Boiling point: 100degreeC, Flash point: >100degreeC, pH: 6.4 at 20degreeC, Ignition temperature: 400.0degreeC