Learn More
Bio Rad Laboratories 4–15% Criterion™ TGX™ Precast Midi Protein Gel, Prep+2 well, 700 µl #5671082
Catalog No. NC3659272
Encompass
Pkg of 1, 4–15% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion™ Dodeca™ electrophoresis cells
Catalog No. NC3659272 Supplier Bio Rad Laboratories Supplier No. 5671082
Pkg of 1, 4–15% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion™ Dodeca™ electrophoresis cells