Learn More
Cell Signaling Technology 4-1BB/CD137/TNFRSF9 D2Z4Y Ra
SDP
Catalog No. 502727038
Encompass
4-1BB/CD137/TNFRSF9 (D2Z4Y) Rabbit mAb 20 ul
Catalog No. 50-272-7038 Supplier Cell Signaling Technology Supplier No. 34594T
Add to Cart
Add to Cart
4-1BB/CD137/TNFRSF9 (D2Z4Y) Rabbit mAb 20 ul