Learn More
ACROBiosystems ACROBiosystems Mouse 4-1BB / TNFRSF9 Protein, His Tag
Supplier:  ACROBiosystems 41BM52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse 4-1BB, His Tag (41B-M52H7) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession # NP_001070976.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) at 0.5 μg/mL (100 μL/well) can bind Mouse 4‑1BB Ligand, Fc Tag (Cat. No. 41L-M5257) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-7306