Learn More
ACROBiosystems ACROBiosystems Rabbit 4-1BB / TNFRSF9 Protein, His Tag
Supplier:  ACROBiosystems 41BR52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Rabbit 4-1BB, His Tag (41B-R52H9) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Ile 191 (Accession # G1SHD7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.8 kDa. The protein migrates as 27-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) at 2 μg/mL (100 μL/well) can bind Rabbit 4-1BB, His Tag (Cat. No. 41B-R52H9) with a linear range of 1-39 ng/mL (QC tested).
Catalog No. 50-195-6785