Learn More
Medchemexpress LLC 4-1BB/TNFRSF9 protein, mouse (C-terminal human Fc fusion) | >95.0% | 10 UG
SDP
Catalog No. 5000292734
Encompass_Preferred
Recombinant mouse 4-1BB/TNFRSF9 extracellular domain fused to a C-terminal human Fc, expressed in HEK293 cells and supplied as a lyophilized protein. Formulated in 20 mM MES, 150 mM NaCl, pH 5.5, the construct corresponds to UniProt P20334 (V24-L187) with an approximate molecular weight of 58.72 kDa. Purity is greater than 95% by reducing SDS-PAGE; store at -20°C and follow the COA for reconstitution and handling.
Catalog No. 50-002-92734 Supplier Medchemexpress LLC Supplier No. HYP7447A10UG
Add to Cart
Add to Cart
Recombinant mouse 4-1BB/TNFRSF9 extracellular domain fused to a C-terminal human Fc, expressed in HEK293 cells and supplied as a lyophilized protein. Formulated in 20 mM MES, 150 mM NaCl, pH 5.5, the construct corresponds to UniProt P20334 (V24-L187) with an approximate molecular weight of 58.72 kDa. Purity is greater than 95% by reducing SDS-PAGE; store at -20°C and follow the COA for reconstitution and handling.