Learn More
Medchemexpress LLC 4-1BB/TNFRSF9 protein, mouse (HEK293, C-hFc) | ≥95.0% | 100 UG
SDP
Catalog No. 5000293828
Encompass_Preferred
Recombinant mouse 4-1BB (TNFRSF9) extracellular domain fused to a C-terminal human Fc, produced in HEK293 cells and supplied as a lyophilized powder for research use. The protein is sequence-verified (V24-L187), has an approximate molecular weight of ~58.7 kDa, and is characterized by high purity and demonstrated functional activity in cell-based assays.
  • Recombinant extracellular domain produced in mammalian (HEK293) expression system.
  • C-terminal human Fc fusion for improved stability and detection.
  • Lyophilized format with reconstitution guidance (do not reconstitute below 100 μg/mL).
  • High purity: ≥95.0% by reducing SDS-PAGE.
  • Approximate molecular weight: 58.72 kDa.
  • Functionally validated: blocks 4-1BBL-induced IL-2 secretion in CTLL-2 cells.
Catalog No. 50-002-93828 Supplier Medchemexpress LLC Supplier No. HYP7447A100UG
Add to Cart
Add to Cart
Recombinant mouse 4-1BB (TNFRSF9) extracellular domain fused to a C-terminal human Fc, produced in HEK293 cells and supplied as a lyophilized powder for research use. The protein is sequence-verified (V24-L187), has an approximate molecular weight of ~58.7 kDa, and is characterized by high purity and demonstrated functional activity in cell-based assays.
  • Recombinant extracellular domain produced in mammalian (HEK293) expression system.
  • C-terminal human Fc fusion for improved stability and detection.
  • Lyophilized format with reconstitution guidance (do not reconstitute below 100 μg/mL).
  • High purity: ≥95.0% by reducing SDS-PAGE.
  • Approximate molecular weight: 58.72 kDa.
  • Functionally validated: blocks 4-1BBL-induced IL-2 secretion in CTLL-2 cells.