Learn More
Genscript Corporation 4-1BB/TNFRSF9Biotin His A
Catalog No. 5000058594
Encompass
4-1BB is also known as CD137 is a type 2 transmembrane glycoprotein receptor belonging to the TNF superfamily CD137 can be expressed by activated T cells but to a larger extent on CD8 than on CD4 T cells In addition CD137 expression is found on dendritic cells B cells follicular dendritic cells natural killer cells granulocytes and cells of blood vessel walls at sites of inflammation
Catalog No. 50-000-58594 Supplier Genscript Corporation Supplier No. Z04385500
4-1BB is also known as CD137 is a type 2 transmembrane glycoprotein receptor belonging to the TNF superfamily CD137 can be expressed by activated T cells but to a larger extent on CD8 than on CD4 T cells In addition CD137 expression is found on dendritic cells B cells follicular dendritic cells natural killer cells granulocytes and cells of blood vessel walls at sites of inflammation