Learn More
Medchemexpress LLC 4-1BBL/TNFSF9 protein, rat (HEK293, His) | >95.0% | 10 UG
SDP
Catalog No. 5000293588
Encompass_Preferred
Recombinant rat 4-1BBL/TNFSF9 extracellular domain expressed in HEK293 cells with a His affinity tag. Designed for receptor-binding and functional immunology assays, the product is reported to span residues P106-G308 (accession Q80WE6) and is characterized for purity and binding activity per the supplier's analytical documents.
  • Mammalian expression preserves native glycosylation and folding.
  • His-tag enables affinity purification and convenient detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Reported sequence covers residues P106-G308 (accession Q80WE6).
  • Validated binding activity in functional ELISA assays.
  • Provided in a small-format quantity suitable for pilot or mechanistic studies.
Catalog No. 50-002-93588 Supplier Medchemexpress LLC Supplier No. HYP7670510UG
Add to Cart
Add to Cart
Recombinant rat 4-1BBL/TNFSF9 extracellular domain expressed in HEK293 cells with a His affinity tag. Designed for receptor-binding and functional immunology assays, the product is reported to span residues P106-G308 (accession Q80WE6) and is characterized for purity and binding activity per the supplier's analytical documents.
  • Mammalian expression preserves native glycosylation and folding.
  • His-tag enables affinity purification and convenient detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Reported sequence covers residues P106-G308 (accession Q80WE6).
  • Validated binding activity in functional ELISA assays.
  • Provided in a small-format quantity suitable for pilot or mechanistic studies.