Learn More
ACROBiosystems ACROBiosystems Mouse 4-1BB Ligand / TNFSF9 Protein, Fc Tag
Catalog No. 501957274
Encompass_Preferred
Recombinant Protein;1MG;Mouse 4-1BB Ligand, Fc Tag (41L-M5257) is expressed from human 293 cells (HEK293). It contains AA Arg 104 - Glu 309 (Accession # NP_033430).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 64-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) at 0.5 μg/mL (100 μL/well) can bind Mouse 4‑1BB Ligand, Fc Tag (Cat. No. 41L-M5257) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-195-7274 Supplier ACROBiosystems Supplier No. 41LM52571MG
Recombinant Protein;1MG;Mouse 4-1BB Ligand, Fc Tag (41L-M5257) is expressed from human 293 cells (HEK293). It contains AA Arg 104 - Glu 309 (Accession # NP_033430).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 64-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) at 0.5 μg/mL (100 μL/well) can bind Mouse 4‑1BB Ligand, Fc Tag (Cat. No. 41L-M5257) with a linear range of 0.4-6 ng/mL (QC tested).