Learn More
ACROBiosystems ACROBiosystems Human 4-1BB Ligand / TNFSF9 (71-254) Protein, His,Flag Tag (active trimer) (MALS verified)
Catalog No. 501956436
Encompass_Preferred
Recombinant Protein;1MG;Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (41L-H52D4) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession # P41273-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 62.8 kDa. The protein migrates as 55-60 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (Cat. No. 41L-H52D4) at 2 μg/mL (100 μL/well) can bind Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-195-6436 Supplier ACROBiosystems Supplier No. 41LH52D41MG
Recombinant Protein;1MG;Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (41L-H52D4) is expressed from human 293 cells (HEK293). It contains AA Arg 71 - Glu 254 (Accession # P41273-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 62.8 kDa. The protein migrates as 55-60 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (Cat. No. 41L-H52D4) at 2 μg/mL (100 μL/well) can bind Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) with a linear range of 0.2-3 ng/mL (QC tested).