Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human 4-1BB Ligand / TNFSF9 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;41L,4-1BB-L,CD137L,4-1BB ligand,4-1BBL,TNFSF9,TNFSF-9,41BB
Catalog No. 501658081
Encompass_Preferred
Recombinant Protein;Biotinylated Human 4-1BB Ligand / TNFSF9 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # P41273-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) at 0.1 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-8081 Supplier ACROBiosystems Supplier No. 41LH82F925UG
Recombinant Protein;Biotinylated Human 4-1BB Ligand / TNFSF9 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (41L-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 50 - Glu 254 (Accession # P41273-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.2 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) at 0.1 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).