Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(1H-1,2,4-Triazol-1-yl)aniline

Catalog No. CDS013675 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013675 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013675 Supplier Sigma Aldrich Supplier No. CDS013675500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Preparation of tetramic acids via dieckmann ring closure
  • Organocatalytic decarboxylative doebner-knoevenagel reactions
  • Conjugation to amino-modified oligonucleotides
  • Acylation reactions (acylation agent)
  • Indole glyoxylamides for potential prion disease treatment
  • Knoevenagel condensation with aldehydestandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. 445088.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 6523-49-5
Molecular Formula C8H8N4
Linear Formula C8H8N4
MDL Number MFCD00171470
Quantity 5 g
Molecular Weight (g/mol) 160.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.