Learn More
Sigma Aldrich 4-(1H-1,3-benzimidazol-1-yl)benzenecarbaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- (2-iodoethyl)benzene is a halogenated hydrocarbon
- It is reported to undergo triethyl borane-mediated intermolecular radical addition with 2h-azirine-3-carboxylate.(2-iodoethyl)benzene is suitable for use as starting reagent in the preparation of organic-inorganic hybrid compounds such as, [c6h5ch2nh3]2pbi4, [c6h5ch2ch2sc(nh2 )2]3pbi5 and [c10h7ch2nh3]pbi3
- It may be used in the preparation of dioxane-based antiviral agents
Specifications
Specifications
| CAS | 90514-72-0 |
| Molecular Formula | C14H10N2O |
| Linear Formula | C14H10N2O |
| MDL Number | MFCD03425762 |
| Quantity | 5 mL |
| Molecular Weight (g/mol) | 222.24 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.