Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1h-imidazol-1-ylmethyl)benzoic acid hydrochloride, Thermo Scientific™

Catalog No. CC43351DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC43351DA 1 g Amber Glass Bottle
CC43351CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC43351DA Supplier Thermo Scientific Chemicals Supplier No. CC43351DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 135611-32-4
Molecular Formula C11H11ClN2O2
Molecular Weight (g/mol) 238.671
MDL Number MFCD08271922
InChI Key MZTHHEXWAYFTBJ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-ylmethyl benzoic acid hydrochloride, 4-1h-imidazol-1-yl methyl benzoic acid hydrochloride, 4-imidazol-1-ylmethyl benzoic acid hydrochloride, acmc-1bx8o, 4-imidazolylmethyl benzoic acid, chloride, 4-1h-imidazol-1-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 18525764
IUPAC Name 4-(imidazol-1-ylmethyl)benzoic acid;hydrochloride
SMILES C1=CC(=CC=C1CN2C=CN=C2)C(=O)O.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.67
Percent Purity 95%
Chemical Name or Material 4-(1H-imidazol-1-ylmethyl)benzoic acid hydrochloride
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.