Learn More
Medchemexpress LLC 4-(1H-indol-4-yl)-N-phenylpiperazine-1-carboxamide | 882256-55-5 | MFCD00830676 | 99.8% | 320.39 g/mol | C19H20N4O | 10MG
SDP
Catalog No. 5000235094
Encompass_Preferred
iGOT1-01 is a small-molecule inhibitor of glutamate oxaloacetate transaminase 1 (GOT1) intended for biochemical and cellular research. The compound is supplied as a white to off-white solid with molecular formula C19H20N4O and a molecular weight of approximately 320.4 g/mol; manufacturer-reported purity is about 99.8%. Recommended formulations and storage guidance are provided for in vitro and in vivo use.
Catalog No. 50-002-35094 Supplier Medchemexpress LLC Supplier No. HY12482510MG
iGOT1-01 is a small-molecule inhibitor of glutamate oxaloacetate transaminase 1 (GOT1) intended for biochemical and cellular research. The compound is supplied as a white to off-white solid with molecular formula C19H20N4O and a molecular weight of approximately 320.4 g/mol; manufacturer-reported purity is about 99.8%. Recommended formulations and storage guidance are provided for in vitro and in vivo use.