Learn More
Medchemexpress LLC 4-(1H-indol-4-yl)-N-phenylpiperazine-1-carboxamide | 882256-55-5 | MFCD00830676 | 99.8% | 320.39 g/mol | C19H20N4O | 10MM 1ML
SDP
Catalog No. 5000235825
Encompass_Preferred
iGOT1-01 is a small-molecule inhibitor of aspartate aminotransferase 1 (GOT1) intended for research use. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as a solid in various sizes. Key identifiers include CAS 882256-55-5, molecular formula C19H20N4O, and molecular weight ~320.4 g/mol. Reported purity is approximately 99.77%.
  • Potent GOT1 inhibitor suitable for biochemical and cell-based assays.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid for custom formulation and storage.
  • High reported purity suitable for research applications.
  • Stable under recommended storage conditions for powder and solvent forms.
  • Contains CAS 882256-55-5 for unambiguous identification.
Catalog No. 50-002-35825 Supplier Medchemexpress LLC Supplier No. HY12482510MM 1ML
iGOT1-01 is a small-molecule inhibitor of aspartate aminotransferase 1 (GOT1) intended for research use. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available as a solid in various sizes. Key identifiers include CAS 882256-55-5, molecular formula C19H20N4O, and molecular weight ~320.4 g/mol. Reported purity is approximately 99.77%.
  • Potent GOT1 inhibitor suitable for biochemical and cell-based assays.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid for custom formulation and storage.
  • High reported purity suitable for research applications.
  • Stable under recommended storage conditions for powder and solvent forms.
  • Contains CAS 882256-55-5 for unambiguous identification.