Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol 97+%, Thermo Scientific™

Catalog No. CC56009CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56009CB 250 mg Amber Glass Bottle
CC56009DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC56009CB Supplier Thermo Scientific Chemicals Supplier No. CC56009CB
Add to Cart

Chemical Identifiers

CAS 160388-55-6
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD08435916
InChI Key ADLULGOPZPFWPA-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl phenyl methanol, 4-pyrazol-1-ylmethyl phenyl methanol, 4-1h-pyrazol-1-yl methyl phenyl methanol, 1-4-hydroxymethyl-benzyl-1h-pyrazole, benzenemethanol, 4-1h-pyrazol-1-ylmethyl, 9vq
PubChem CID 16640562
IUPAC Name [4-(pyrazol-1-ylmethyl)phenyl]methanol
SMILES OCC1=CC=C(CN2C=CC=N2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material [4-(1H-Pyrazol-1-ylmethyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.