Learn More
Medchemexpress LLC 4(1H)-Pyrimidinone, 2,3-dihydro-2-selenoxo- | 16724-03-1 | 98.0% | 175.05 | 100 MG
SDP
Catalog No. 5000439275
Encompass_Preferred
2-Selenouracil is a specialized photosensitizer useful for photodynamical therapy. This compound exhibits efficient population of triplet states while maintaining photostability, as highlighted in recent research.
  • Functions as a specialized photosensitizer
  • Applicable for photodynamical therapy
  • Exhibits efficient triplet state population
  • Maintains photostability
Catalog No. 50-004-39275 Supplier Medchemexpress LLC Supplier No. HY111959100MG
2-Selenouracil is a specialized photosensitizer useful for photodynamical therapy. This compound exhibits efficient population of triplet states while maintaining photostability, as highlighted in recent research.
  • Functions as a specialized photosensitizer
  • Applicable for photodynamical therapy
  • Exhibits efficient triplet state population
  • Maintains photostability