Learn More
Medchemexpress LLC 4-[(1R)-1-aminoethyl]-N-(1H-pyrrolo[2,3-b]pyridin-4-yl)benzamide | 199433-58-4 | 280.32 g/mol | C16H16N4O | 50 MG
SDP
Catalog No. 5000213828
Encompass_Preferred
Y-33075 is a potent, selective Rho-associated protein kinase (ROCK) inhibitor intended for research use. It shows high biochemical potency with a reported IC50 of approximately 3.6 nM and is used to study ROCK signaling in both enzymatic and cellular assays.
  • Potent and selective ROCK inhibition with IC50 ≈ 3.6 nM.
  • Derived from earlier ROCK inhibitors, offering improved potency.
  • Suitable for biochemical and cellular studies of ROCK signaling.
  • Chemical formula C16H16N4O and molar mass 280.32 g/mol.
  • Supplied in small research pack sizes appropriate for assay development.
Catalog No. 50-002-13828 Supplier Medchemexpress LLC Supplier No. HY1006750MG
Y-33075 is a potent, selective Rho-associated protein kinase (ROCK) inhibitor intended for research use. It shows high biochemical potency with a reported IC50 of approximately 3.6 nM and is used to study ROCK signaling in both enzymatic and cellular assays.
  • Potent and selective ROCK inhibition with IC50 ≈ 3.6 nM.
  • Derived from earlier ROCK inhibitors, offering improved potency.
  • Suitable for biochemical and cellular studies of ROCK signaling.
  • Chemical formula C16H16N4O and molar mass 280.32 g/mol.
  • Supplied in small research pack sizes appropriate for assay development.