Learn More
Medchemexpress LLC 4-[(1R)-1-aminoethyl]-N-(1H-pyrrolo[2,3-b]pyridin-4-yl)benzamide | 199433-58-4 | >98.0% | 280.33 g/mol | C16H16N4O | 5 MG
SDP
Catalog No. 5000214189
Encompass_Preferred
This small-molecule Rho-associated kinase (ROCK) inhibitor is derived from Y-27632 and demonstrates potent ROCK inhibition (IC50 3.6 nM). It also shows inhibitory activity against PKC (IC50 ≈420 nM) and CaMKII (IC50 ≈810 nM). Supplied as a solid for research use, it is intended for in vitro studies of cytoskeletal dynamics, cell morphology, and signaling pathways.
  • Potent ROCK inhibition (IC50 3.6 nM).
  • Activity against PKC and CaMKII at higher concentrations.
  • Solid form supplied for laboratory research use.
  • Suitable for studies of cytoskeletal regulation and cell motility.
  • Available in small pre-weighed pack sizes for experimental flexibility.
Catalog No. 50-002-14189 Supplier Medchemexpress LLC Supplier No. HY100675MG
This small-molecule Rho-associated kinase (ROCK) inhibitor is derived from Y-27632 and demonstrates potent ROCK inhibition (IC50 3.6 nM). It also shows inhibitory activity against PKC (IC50 ≈420 nM) and CaMKII (IC50 ≈810 nM). Supplied as a solid for research use, it is intended for in vitro studies of cytoskeletal dynamics, cell morphology, and signaling pathways.
  • Potent ROCK inhibition (IC50 3.6 nM).
  • Activity against PKC and CaMKII at higher concentrations.
  • Solid form supplied for laboratory research use.
  • Suitable for studies of cytoskeletal regulation and cell motility.
  • Available in small pre-weighed pack sizes for experimental flexibility.