Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine 95.0+%, TCI America™
SDP

Catalog No. T35971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35971G 1 g
T35975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35971G Supplier TCI America Supplier No. T35971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 289706-46-3
Molecular Formula C9H18F3NO2Si

Specifications

Color Yellow
Quantity 1 g
Formula Weight 257.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.