Learn More
Cayman Chemical 4-(2,2,3-Trimethylcyclopentyl)butanoic Acid, Purity: ≥98%, CAS: 957136-80-0, 50 mg
SDP
Catalog No. 502983445
Encompass_Preferred
An orthosteric, insurmountable antagonist of the human bitter taste receptors hTAS2R31 and hTAS2R43; prevents the activation of hTAS2R31 by saccharin and acesulfame K in cells (IC50s = 7.9 and 2.4 µM, respectively)
Catalog No. 50-298-3445 Supplier Cayman Chemical Supplier No. 1960650 MG
An orthosteric, insurmountable antagonist of the human bitter taste receptors hTAS2R31 and hTAS2R43; prevents the activation of hTAS2R31 by saccharin and acesulfame K in cells (IC50s = 7.9 and 2.4 µM, respectively)