Learn More
Medchemexpress LLC 4-[2-[(2,4,6-trifluorophenoxy)methyl]phenyl]piperidine hydrochloride | 1227056-87-2 | 99.5% | 357.80 | C18H19ClF3NO | 50 MG
SDP
Catalog No. 5000240014
Encompass_Preferred
Ampreloxetine hydrochloride is the hydrochloride salt of a small-molecule norepinephrine and serotonin reuptake inhibitor used for research into NET/SERT pharmacology and central nervous system activity. Supplied as a dry powder for laboratory use, it is intended for in vitro and preclinical studies.
  • Hydrochloride salt form for improved stability.
  • Potent norepinephrine and serotonin reuptake inhibition.
  • Suitable for in vitro and preclinical research.
  • Dry powder form for accurate dosing and formulation.
  • High reported purity.
  • Characterized by CAS 1227056-87-2 and molecular weight 357.80.
Catalog No. 50-002-40014 Supplier Medchemexpress LLC Supplier No. HY10712850MG
Ampreloxetine hydrochloride is the hydrochloride salt of a small-molecule norepinephrine and serotonin reuptake inhibitor used for research into NET/SERT pharmacology and central nervous system activity. Supplied as a dry powder for laboratory use, it is intended for in vitro and preclinical studies.
  • Hydrochloride salt form for improved stability.
  • Potent norepinephrine and serotonin reuptake inhibition.
  • Suitable for in vitro and preclinical research.
  • Dry powder form for accurate dosing and formulation.
  • High reported purity.
  • Characterized by CAS 1227056-87-2 and molecular weight 357.80.