Learn More
Medchemexpress LLC 4-(2,2-difluoro-1,3-benzodioxol-4-yl)-1H-pyrrole-3-carbonitrile | 131341-86-1 | MFCD01631158 | 98.0% | 248.19 g/mol | C12H6F2N2O2 | 200 MG
SDP
Catalog No. 5000300761
Encompass_Preferred
Fludioxonil is a phenylpyrrole fungicide supplied as an analytical standard (CAS 131341-86-1) for laboratory research and method development. The material is provided as a high-purity solid suitable for reference, calibration, and analytical assays.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular formula C12H6F2N2O2 and molecular weight 248.19 g/mol.
  • Supplied as a stable solid for reliable storage and handling.
  • Suitable for method development, calibration, and reference standards.
  • Compatible with HPLC and GC analytical techniques.
Catalog No. 50-003-00761 Supplier Medchemexpress LLC Supplier No. HYW021040200MG
Fludioxonil is a phenylpyrrole fungicide supplied as an analytical standard (CAS 131341-86-1) for laboratory research and method development. The material is provided as a high-purity solid suitable for reference, calibration, and analytical assays.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular formula C12H6F2N2O2 and molecular weight 248.19 g/mol.
  • Supplied as a stable solid for reliable storage and handling.
  • Suitable for method development, calibration, and reference standards.
  • Compatible with HPLC and GC analytical techniques.