Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline 98.0+%, TCI America™
SDP

Catalog No. D44785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44785G 5 g
D447825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44785G Supplier TCI America Supplier No. D44785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89114-91-0
Molecular Formula C34H29N
Molecular Weight (g/mol) 451.613
MDL Number MFCD09832972
InChI Key ZHGLWMUJQVWWQO-UHFFFAOYSA-N
PubChem CID 14549579
IUPAC Name N-[4-(2,2-diphenylethenyl)phenyl]-4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Quantity 5 g
Formula Weight 451.61
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,2-Diphenylethenyl)-N,N-di(p-tolyl)aniline
EINECSNumber (4)-1576
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.