Learn More
Medchemexpress LLC 4-[2-(2-fluorophenyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl]pyridine | 1784751-19-4 | MFCD29924749 | 99.9% | 333.33 g/mol | C20H13F2N3 | 50 MG
SDP
Catalog No. 5000215151
Encompass_Preferred
TA-02 is a small-molecule p38 MAPK inhibitor used in biochemical and cell-based research. As an analog of SB 203580, it exhibits potent activity (IC50 ≈ 20 nM) and preferential inhibition of TGFBR-2. The compound is supplied as a high-purity solid suitable for laboratory studies involving kinase signaling modulation.
  • Potent p38 MAPK inhibition (IC50 ≈ 20 nM).
  • Preferential inhibition of TGFBR-2.
  • High purity solid suitable for research applications.
  • Compact pack sizes available for small-scale studies.
  • Molecular weight ~333.33 g/mol; formula C20H13F2N3.
Catalog No. 50-002-15151 Supplier Medchemexpress LLC Supplier No. HY10011550MG
TA-02 is a small-molecule p38 MAPK inhibitor used in biochemical and cell-based research. As an analog of SB 203580, it exhibits potent activity (IC50 ≈ 20 nM) and preferential inhibition of TGFBR-2. The compound is supplied as a high-purity solid suitable for laboratory studies involving kinase signaling modulation.
  • Potent p38 MAPK inhibition (IC50 ≈ 20 nM).
  • Preferential inhibition of TGFBR-2.
  • High purity solid suitable for research applications.
  • Compact pack sizes available for small-scale studies.
  • Molecular weight ~333.33 g/mol; formula C20H13F2N3.